AS-252424 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

A129503-100MG A129503-1MG A129503-25MG A129503-2MG A129503-5MG
ALA129503100MGEA 1401.16 USD
ALA129503100MG ALA129503-1MG ALA129503-25MG ALA129503-2MG ALA129503-5MG
AS-252424 ≥98%, Moligand™
AS-252424

AS-252424 is a novel, potent PI3Kγ inhibitor with IC50 of 30 nM with 30-fold selectivity for PI3Kγ than PI3Kα, and low inhibitory activity towards PI3Kδ/β.A selective PI 3-kinase p110γ inhibitor.

Formula: C₁₄H₈FNO₄S
MW: 305.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045281
CAS Number: 900515-16-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0cf40c0ad2fbf1f1381a08929d7de020688122&vid=ea0cf40c0ad2fbf1f1381a08929d7de020688122&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR