Amprolium hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A129501-100G A129501-1G A129501-25G A129501-5G
ALA129501-100GEA 205.11 USD
ALA129501-100G ALA129501-1G ALA129501-25G ALA129501-5G
Amprolium hydrochloride ≥98%
Amprolium hydrochloride

Amprolium chloride is a thiamin antagonist, which prevents carbohydrate synthesis by blocking thiamine uptakeThiamine analog coccidiostat.

Formula: C₁₄H₁₉ClN₄
Melting Pt: 241 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078831
CAS Number: 137-88-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ea0e8b49b3431779d83068f41115af89c6b46e6&vid=4ea0e8b49b3431779d83068f41115af89c6b46e6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR