Securopen ≥99%

Supplier: ALADDIN SCIENTIFIC
Danger

A129398-1G A129398-25G A129398-5G
ALA129398-1GEA 109.16 USD
ALA129398-1G ALA129398-25G ALA129398-5G
Securopen ≥99%
Securopen

Azlocillin is an acylampicillin with a broad spectrum against bacteria.Azlocillin is a semisynthetic penicillin and is a potent 1-lactam antibiotic. It is often used against Pseudomonas aeruginosa and has been used to develop novel antibiotic quantitation techniques

Formula: C₂₀H₂₃N₅NaO₆S
Storage Temperature: Refrigerator
MDL Number: MFCD07793330
CAS Number: 37091-65-9
EINECS: 253-347-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b6a680b74d03fabc4efa1281959f36bf4981233&vid=5b6a680b74d03fabc4efa1281959f36bf4981233&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR