Adiphenine hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Spasnil, Diphenylacetic Acid 2-(Diethylamino)ethyl Ester Hydrochloride, Adiphenine HCl, Diphenylacetic acid 2-(Diethylamino)ethyl Ester Hydrochloride, Adiphenine Hydrochloride, Trasentin Hydrochloride, 2-(Diethylamino)ethyl Diphenylacetate Hydrochloride, Patrovina, Sentiv

A129364-100G A129364-10G A129364-25G A129364-50G A129364-5G
ALA129364-100GEA 257.36 USD
ALA129364-100G ALA129364-10G ALA129364-25G ALA129364-50G ALA129364-5G
Adiphenine hydrochloride ≥98%
Adiphenine hydrochloride

Adiphenine HCl is a nicotinic receptor inhibitor with IC50 of 15 μM.

Formula: C₂₀H₂₅NO₂
MW: 347.88 g/mol
Melting Pt: 117 °C
Storage Temperature: Freezer
MDL Number: MFCD00050508
CAS Number: 50-42-0
EINECS: 200-036-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f273c954bf5e8739fd91ed63988772a45c03064d&vid=f273c954bf5e8739fd91ed63988772a45c03064d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR