Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulfate ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
Atazanavir sulphate
A129321-1G
A129321-250MG
A129321-50MG
A129321-5G
ALA129321-1GEA
2401.51
USD
ALA129321-1G
ALA129321250MG
ALA129321-50MG
ALA129321-5G
Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulfate ≥98%
Methyl ((5S,10S,11S,14S)-11-benzyl-5-(tert-butyl)-10-hydroxy-15,15-dimethyl-3,6,13-trioxo-8-(4-(pyridin-2-yl)benzyl)-2-oxa-4,7,8,12-tetraazahexadecan-14-yl)carbamate sulfate
Atazanavir is a HIV protease inhibitor with Ki of 2.66 nM.A sulfate salt form of atazanavir that is an highly potent HIV-1 protease inhibitor.
Formula:
C₃₈H₅₄N₆O₁₁S MW: 802.93 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08067748 CAS Number: 229975-97-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...