11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Azatadine dimaleate

A129250-10MG A129250-1G A129250-250MG A129250-50MG
ALA129250-10MGEA 55.96 USD
ALA129250-10MG ALA129250-1G ALA129250250MG ALA129250-50MG
11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate ≥98%
11-(1-Methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine dimaleate
Formula: C₂₈H₃₀N₂O₈
MW: 522.54 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00242593
CAS Number: 3978-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c464ee5d0fbd2f4d038edd829cc54149a343b60&vid=0c464ee5d0fbd2f4d038edd829cc54149a343b60&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR