Aliskiren hemifumarate ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A129231-500MG A129231-5G A129231-100MG A129231-1G A129231-250MG
ALA129231500MGEA 904.31 USD
ALA129231500MG ALA129231-5G ALA129231100MG ALA129231-1G ALA129231250MG
Aliskiren hemifumarate ≥98%
Aliskiren hemifumarate

Aliskiren hemifumarate is a direct renin inhibitor with IC50 of 1.5 nM.

Formula: C₃₀H₅₃N₃O₆·0.5C₄H₄O₄
Storage Temperature: Freezer
MDL Number: MFCD27756642
CAS Number: 173334-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5d80fde38761af5b9dfcc213e3ad02766828816&vid=b5d80fde38761af5b9dfcc213e3ad02766828816&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR