3-Aminobenzene sulfonic acid ≥98%
Supplier: Aladdin Scientific
|
Warning
|
A129175-100G
A129175-25G
ALA129175-100GEA
22.71
USD
ALA129175-100G
ALA129175-25G
3-Aminobenzene sulfonic acid ≥98%
3-Aminobenzene sulphonic acid
3-Aminobenzenesulfonic acid was used in the synthesis of cytocompatible sulfonated polyanilines. It was used in fabrication of novel glucose biosensor having large active surface area and excellent conductivity.
Formula:
C₆H₇NO₃S Melting Pt: >300°C |
MDL Number:
MFCD00065345 CAS Number: 121-47-1 EINECS: 204-473-6 UN: 2585 ADR: 8,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...