4-Aminonicotinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-3-pyridinecarboxylic acid, 4-Amino-nicotinic acid, 4-Amino-3-pyridinecarboxylic acid

A129122-1G A129122-5G
ALA129122-1GEA 10.36 USD
ALA129122-1G ALA129122-5G
4-Aminonicotinic acid ≥98%
4-Aminonicotinic acid

4-Amino-nicotinic acid is a white to light yellow and faint beige to light beige and faint brown to light brown powder or crystals.4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine.

Formula: C₆H₆N₂O₂
Melting Pt: 307-312°C
MDL Number: MFCD00234183
CAS Number: 7418-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10f1cfae54a7d4713cd06207207b3603d3108b1&vid=a10f1cfae54a7d4713cd06207207b3603d3108b1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR