Aminoacetaldehyde diethyl acetal ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal

A129111-25ML A129111-5ML
ALA129111-25MLEA 65.46 USD
ALA129111-25ML ALA129111-5ML
Aminoacetaldehyde diethyl acetal ≥97%
Aminoacetaldehyde diethyl acetal
Formula: C₆H₁₅NO₂
Boiling Pt: 162-163°C
Density: 0.916 g/cm³
Flash Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00008136
CAS Number: 645-36-3
EINECS: 211-439-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae27ed65dc6ca14a0329a212d2629744259f228b&vid=ae27ed65dc6ca14a0329a212d2629744259f228b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR