2-Aminoimidazole hemisulfate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)

A129107-100G A129107-1G A129107-25G A129107-5G
ALA129107-100GEA 790.31 USD
ALA129107-100G ALA129107-1G ALA129107-25G ALA129107-5G
2-Aminoimidazole hemisulfate ≥98%
2-Aminoimidazole hemisulphate

2-Aminoimidazole sulfate was used in the synthesis of chlorohydrin

Formula: C₆H₁₂N₆O₄S
MW: 264.26 g/mol
Melting Pt: 270°C
MDL Number: MFCD00013162
CAS Number: 1450-93-7
EINECS: 215-918-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e198b58f105106bd120b9447e354f503970f5a50&vid=e198b58f105106bd120b9447e354f503970f5a50&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR