2-Amino-5-iodopyridine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

A129106-1G A129106-25G A129106-5G
ALA129106-1GEA 24.61 USD
ALA129106-1G ALA129106-25G ALA129106-5G
2-Amino-5-iodopyridine ≥98%
2-Amino-5-iodopyridine
Formula: C₅H₅IN₂
MW: 220.01
Boiling Pt: 271
Melting Pt: 128-131°C
Storage Temperature: Refrigerator
MDL Number: MFCD00160312
CAS Number: 20511-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=957d3c1a85d5ac7f668a49bd725235d6a3f44e49&vid=957d3c1a85d5ac7f668a49bd725235d6a3f44e49&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR