2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine, 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
A128909-1G
A128909-250MG
A128909-50MG
ALA128909-1GEA
421.71
USD
ALA128909-1G
ALA128909250MG
ALA128909-50MG
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥95%
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester
Application:2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.
Formula:
C₁₂H₁₆BF₃N₂O₂ MW: 288.07 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12923420 CAS Number: 944401-57-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...