Amlodipine besylate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylic acid 3-Ethyl 5-Methyl Ester Benzenesulphon, 3-Ethyl 5-Methyl 2-(2-Aminoethoxy)methyl-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Benzenesulphonate, 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate benzenesulphonate

A128786-1G A128786-25G
ALA128786-1GEA 55.96 USD
ALA128786-1G ALA128786-25G
Amlodipine besylate ≥98%
Amlodipine besylate

Related CAS Number :88150-42-9Amlodipine besylate is a L-type calcium channel blocker which may be used for the treatment of angina pectoris and hypertension. Amlodipine also inhibits growth of human epidermoid carcinoma A431 cells and has antireproductive effects in male rats.

Formula: C₂₀H₂₅ClN₂O₅
MW: 567.05 g/mol
Melting Pt: 195-204°C
MDL Number: MFCD00887594
CAS Number: 111470-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d64500c66df668b6727f1da77d91bdd5569b7227&vid=d64500c66df668b6727f1da77d91bdd5569b7227&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR