3-Aminopyrrolidine dihydrochloride ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Pyrrolidin-3-amine dihydrochloride
A128215-1G
A128215-25G
A128215-5G
ALA128215-1GEA
17.96
USD
ALA128215-1G
ALA128215-25G
ALA128215-5G
3-Aminopyrrolidine dihydrochloride ≥98%
3-Aminopyrrolidine dihydrochloride
3-Aminopyrrolidine dihydrochloride (3-pyrrolidinamine dihydrochloride) is one of the key intermediate of tosufloxacin and other quinolone antibiotics.3-Aminopyrrolidine dihydrochloride is the suitable reagent used as an internal standard for the quantitative analysis of amino acids in bio-fluids. (R,S) 3-Aminopyrrolidine dihydrochloride may be used in the preparation of cis-[PdCl2(pyrr)] and cis-[PtCl2(pyrr)] (pyrr= (R,S)-3-aminopyrrolidine) complexes.
Formula:
C₄H₁₀N₂ MW: 159.05 g/mol Melting Pt: >300°C Storage Temperature: Ambient |
MDL Number:
MFCD00060176 CAS Number: 103831-11-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...