3-Methylanthranil acid ≥99%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
2-Amino-3-methylbenzoic acid
A128105-100G
A128105-1G
A128105-25G
A128105-5G
ALA128105-100GEA
127.21
USD
ALA128105-100G
ALA128105-1G
ALA128105-25G
ALA128105-5G
3-Methylanthranil acid ≥99%
3-Methylanthranil acid
Used in: 1)Regioselective Suzuki coupling 2)Preparation of inhibitors of 17β-hydroxysteroid dehydrogenase type 1 3)Preparation of boronic esters
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Melting Pt: 174-177°C Storage Temperature: Ambient |
MDL Number:
MFCD00007745 CAS Number: 4389-45-1 EINECS: 224-505-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...