AZD-5363 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, AZD 5363, AZD5363, (S)-4-amino-N-(1-(4-chlorophenyl)-3-hydroxypropyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, Capivasertib, AZD-5363

A128036-100MG A128036-25MG A128036-50MG A128036-5MG
ALA128036100MGEA 564.21 USD
ALA128036100MG ALA128036-25MG ALA128036-50MG ALA128036-5MG
AZD-5363 ≥98%, Moligand™
AZD-5363

AZD5363 potently inhibits all isoforms of Akt(Akt1/Akt2/Akt3) with IC50 of 3 nM/8 nM/8 nM in cell-free assays, similar to P70S6K/PKA and lower activity towards ROCK1/2.

Formula: C₂₁H₂₅ClN₆O₂
Storage Temperature: Freezer
MDL Number: MFCD22628785
CAS Number: 1143532-39-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2afc8fbf98af4fb7cd77c2d6afe1d8706118d7f5&vid=2afc8fbf98af4fb7cd77c2d6afe1d8706118d7f5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR