AR-42 ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A127913-5MG A127913-100MG A127913-10MG A127913-1MG A127913-25MG A127913-50MG
ALA127913-5MGEA 310.56 USD
ALA127913-5MG ALA127913100MG ALA127913-10MG ALA127913-1MG ALA127913-25MG ALA127913-50MG
AR-42 ≥98%
AR-42
Formula: C₁₈H₂₀N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD17676151
CAS Number: 935881-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b075f2a854fb29ca624752adaed40e6c1e595c2&vid=1b075f2a854fb29ca624752adaed40e6c1e595c2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR