Apatinib mesylate ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Apatinib, N-[4-(1-Cyanocyclopentyl)phenyl]-2-[(4-pyridinylmethyl)amino]-3-pyridinecarboxamide methanesulphonate, YN-968D1, N-(4-(1-cyanocyclopentyl)phenyl)-2-((4-methylpyridin-3-yl)amino)nicotinamide methanesulphonate, YN 968D1, YN968D1, Protein Tyrosine Kinase Inhibitors

A127726-100MG A127726-50MG
ALA127726100MGEA 1161.76 USD
ALA127726100MG ALA127726-50MG
Apatinib mesylate ≥99%
Apatinib mesylate

Apatinib(YN-968D1) is an orally bioavailable, selective VEGFR2 inhibitor with IC50 of 1 nM.

Formula: C₂₅H₂₇N₅O₄S
MW: 493.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22420814
CAS Number: 1218779-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02723bc2ac397455adf1cd3a4eab0269d4e08f95&vid=02723bc2ac397455adf1cd3a4eab0269d4e08f95&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR