Ascomycin ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (3S,4R,5S,8R,9E,12S,14S,15R,16S,18R,19R,26aS)-8-Ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-[(1E)-2-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethenyl]-14,16-dimethoxy-4,10,12,18-tetramethyl-15,19-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclotricosine-1,7,20,21(4H,23H)-tetrone

A127632-100MG A127632-1G A127632-25MG A127632-500MG
ALA127632100MGEA 95.86 USD
ALA127632100MG ALA127632-1G ALA127632-25MG ALA127632500MG
Ascomycin ≥97%
Ascomycin

Ascomycin(Immunomycin, FR-900520, FK520) is an ethyl analog of tacrolimus (FK506) with strong immunosuppressant properties.A potent immunosuppressive agentapplication:Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution® is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

Formula: C₄₃H₆₉NO₁₂
MW: 790 g/mol
Boiling Pt: 868℃
Flash Pt: >110°C
Storage Temperature: Freezer
MDL Number: MFCD06198665
CAS Number: 104987-12-4
EINECS: 200-835-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51fefb381d522937b67e931355abbbda07244972&vid=51fefb381d522937b67e931355abbbda07244972&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR