AZD-2014 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: AZD 2014, Vistusertib, AZD-2014, 3-(2,4-bis((3s)-3-methylmorpholin-4-yl)pyrido(5,6-e)pyrimidin-7-yl)-n-methylbenzamide, AZD2014, 3-(2,4-Bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-N-methylbenzamide, 3-(2,4-bis((S)-3-methylmorpholino)pyrido[2,3-d]pyrimidin-7-yl)-N-methylbenzamide

A127600-25MG A127600-5MG
ALA127600-25MGEA 355.21 USD
ALA127600-25MG ALA127600-5MG
AZD-2014 ≥98%, Moligand™
AZD-2014

AZD2014 is a novel mTOR inhibitor with IC50 of 2.8 nM; highly selective against multiple PI3K isoforms (α/β/γ/δ).A selective inhibitor of mTOR kinase

Formula: C₂₅H₃₀N₆O₃
MW: 462.5 g/mol
Density: 1.2 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD22628784
CAS Number: 1009298-59-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49f97070ff738100f4bc8a02083916aed2031b69&vid=49f97070ff738100f4bc8a02083916aed2031b69&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR