ABC294640 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 1S,3R,5R,7S)-3-(4-chlorophenyl)-N-(pyridin-4-ylmethyl)adamantane-1-carboxamide

A127517-10MG A127517-1MG A127517-25MG A127517-50MG A127517-5MG
ALA127517-10MGEA 157.61 USD
ALA127517-10MG ALA127517-1MG ALA127517-25MG ALA127517-50MG ALA127517-5MG
ABC294640 ≥98%, Moligand™
ABC294640
Formula: C₂₃H₂₅ClN₂O
MW: 380.91 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22666355
CAS Number: 915385-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89c1a6b5b0293a13912cb7b00b2491c7844c8b62&vid=89c1a6b5b0293a13912cb7b00b2491c7844c8b62&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR