AT9283 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1-Cyclopropyl-3-(3-(5-(morpholinomethyl)-1H-benzo[d]imidazol-2-yl)-1H-pyrazol-4-yl)urea

A127488-50MG A127488-5MG A127488-10MG A127488-25MG
ALA127488-50MGEA 828.31 USD
ALA127488-50MG ALA127488-5MG ALA127488-10MG ALA127488-25MG
AT9283 ≥98%, Moligand™
AT9283

AT9283 is a small molecule a multi-targeted inhibitor with IC50s of 4, 1.2, 1.1 and approximate 3 nM for Bcr-Abl(T3151), JAK2 and JAK3, Aurora A and Aurora B, respectively.An inhibitor of Aurora A, Aurora B, JAK3, JAK2, and c-Abl

Formula: C₁₉H₂₃N₇O₂
MW: 381.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12031513
CAS Number: 896466-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11bcbe84a1876c2e7bc731f4957b95e80c61091c&vid=11bcbe84a1876c2e7bc731f4957b95e80c61091c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR