AZD-4547 ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: AZD-4547, AZD4547, N-(5-(3,5-dimethoxyphenethyl)-1H-pyrazol-3-yl)-4-((3S,5R)-3,5-dimethylpiperazin-1-yl)benzamide, rel-N-[5-[2-(3,5-Dimethoxyphenyl)ethyl]-1H-pyrazol-3-yl]-4-[(3R,5S)-3,5-dimethyl-1-piperazinyl]benzamide, AZD 4547, rel-N-(5-(3,5-Dimethoxyphenethyl)-1H-pyrazol-3-yl)-4-((3R,5S)-3,5-dimethylpiperazin-1-yl)benzamide

A127209-100MG A127209-10MG A127209-25MG A127209-50MG A127209-5MG
ALA127209100MGEA 235.51 USD
ALA127209100MG ALA127209-10MG ALA127209-25MG ALA127209-50MG ALA127209-5MG
AZD-4547 ≥99%, Moligand™
AZD-4547

AZD4547 is a novel selective FGFR inhibitor targeting FGFR1/2/3 with IC50 of 0.2 nM/2.5 nM/1.8 nM, weaker activity against FGFR4, VEGFR2(KDR), and little activity observed against IGFR, CDK2, and p38. Phase 2/3.A novel selective FGFR inhibitor targeting FGFR1, FGFR2, and FGFR3.

Formula: C₂₆H₃₃N₅O₃
MW: 463.57 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22580423
CAS Number: 1035270-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85eecaba1f1c743324d6cbb629fdd685d93cf084&vid=85eecaba1f1c743324d6cbb629fdd685d93cf084&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR