Abacavir ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol, ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol

A126552-1G A126552-250MG A126552-5G
ALA126552-1GEA 47.41 USD
ALA126552-1G ALA126552250MG ALA126552-5G
Abacavir ≥98%, Moligand™
Abacavir

Abacavir is a powerful nucleoside analog reverse transcriptase inhibitor (NRTI) used to treat HIV and AIDS. Abacavir exhibits antiviral activity. It is associated with myocardial infarction and fatal hypersensitivity reaction (HSR).

Formula: C₁₄H₁₈N₆O
MW: 286.34 g/mol
Melting Pt: 161 °C
Storage Temperature: Freezer
MDL Number: MFCD00903850
CAS Number: 136470-78-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75ad7da6cc7121282e6b7208f2f8a39dbbc2ba1a&vid=75ad7da6cc7121282e6b7208f2f8a39dbbc2ba1a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR