Anastrozole ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: alpha,alpha,alpha',alpha'-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile

A126470-5G A126470-100MG A126470-1G A126470-25G
ALA126470-5GEA 194.66 USD
ALA126470-5G ALA126470100MG ALA126470-1G ALA126470-25G
Anastrozole ≥99%, Moligand™
Anastrozole

Anastrozole is a potent, highly selective aromatase inhibitor, which inhibits human placental aromatase with an IC50 of 15 nM.

Formula: C₁₇H₁₉N₅
MW: 293.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00866298
CAS Number: 120511-73-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc9b7a14f8994b757079091cd1003f2aabf095c&vid=3fc9b7a14f8994b757079091cd1003f2aabf095c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR