AT-406 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: (5S,8S,10aR)-N-benzhydryl-5-((S)-2-(methylamino)propanamido)-3-(3-methylbutanoyl)-6-oxodecahydropyrrolo[1,2-a][1,5]diazocine-8-carboxamide, AT406, (5S,8S,10aR,Z)-N-benzhydryl-5-((Z)-((S)-1-hydroxy-2-(methylamino)propylidene)amino)-3-(3-methylbutanoyl)-6-oxodecahydropyrrolo[1,2-a][1,5]diazocine-8-carbimidic acid, ARRY-334543, Xevinapant, Debio1143, SM-406

A126412-10MG A126412-25MG A126412-50MG A126412-5MG
ALA126412-10MGEA 154.76 USD
ALA126412-10MG ALA126412-25MG ALA126412-50MG ALA126412-5MG
AT-406 ≥98%, Moligand™
AT-406
Formula: C₃₂H₄₃N₅O₄
MW: 561.73 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22124467
CAS Number: 1071992-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46d35a9a9bd4df433db208e1207392be82608f4a&vid=46d35a9a9bd4df433db208e1207392be82608f4a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR