ARRY-520 ≥98%

Supplier: ALADDIN SCIENTIFIC

A126249-10MG A126249-5MG
ALA126249-10MGEA 153.81 USD
ALA126249-10MG ALA126249-5MG
ARRY-520 ≥98%
ARRY-520
Formula: C₂₀H₂₂F₂N₄O₂S
Storage Temperature: Freezer
CAS Number: 885060-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cb183c5753e1f536996af2b52aa42d12517e761&vid=2cb183c5753e1f536996af2b52aa42d12517e761&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR