A-740003 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: A 740003, N-(1-{[(cyanoimino)(5-quinolinylamino) methyl] amino}-2,2-dimethylpropyl)-2-(3,4-dimethoxyphenyl)acetamide, A740003, N-[1-[[(Cyanoamino)(5-quinolinylimino)methyl]amino]-2,2-dimethylpropyl]-3,4-dimethoxybenzeneacetamide

A126213-100MG A126213-250MG A126213-500MG A126213-50MG
ALA126213100MGEA 102.51 USD
ALA126213100MG ALA126213250MG ALA126213500MG ALA126213-50MG
A-740003 ≥98%, Moligand™
A-740003

A-740003 is potent, selective and competitive P2X7 receptor antagonist (IC50 values are 18 and 40 nM for rat and human receptors respectively).A potent, selective and competitive P2X7 receptor antagonist

Formula: C₂₆H₃₀N₆O₃
MW: 474.55 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09970333
CAS Number: 861393-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c20a57b1e4441853835aac63b87a2eb05a4c3e46&vid=c20a57b1e4441853835aac63b87a2eb05a4c3e46&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR