(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile ≥99%, Moligand™
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
Alogliptin
A126201-1G
A126201-250MG
A126201-5G
ALA126201-1GEA
6387.71
USD
ALA126201-1G
ALA126201250MG
ALA126201-5G
(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile ≥99%, Moligand™
(R)-2-((6-(3-Aminopiperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile
Alogliptin is a potent, selective inhibitor of DPP-4 with IC50 of <10 nM, exhibits greater than 10,000-fold selectivity over DPP-8 and DPP-9.
Formula:
C₁₈H₂₁N₅O₂ MW: 339.39 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD09833196 CAS Number: 850649-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...