tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulfonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Asunaprevir

A126103-100MG A126103-10MG A126103-25MG A126103-50MG
ALA126103100MGEA 1361.26 USD
ALA126103100MG ALA126103-10MG ALA126103-25MG ALA126103-50MG
tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulfonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate ≥98%, Moligand™
tert-Butyl ((S)-1-((2S,4R)-4-((7-chloro-4-methoxyisoquinolin-1-yl)oxy)-2-(((1R,2S)-1-((cyclopropylsulfonyl)carbamoyl)-2-vinylcyclopropyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)carbamate

Asunaprevir (BMS-650032) is a potent hepatitis C virus (HCV) NS3 protease inhibitor.An inhibitor of the viral enzyme serine protease HCV NS3.

Formula: C₃₅H₄₆ClN₅O₉S
MW: 748.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD27987900
CAS Number: 630420-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=589297b98ff1c31ef0abb8586193117599448576&vid=589297b98ff1c31ef0abb8586193117599448576&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR