Atazanavir ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 1,14-Dimethyl (3S,8S,9S,12S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate

A125825-250MG A125825-25G A125825-50MG A125825-5G A125825-10MG A125825-1G
ALA125825250MGEA 18.91 USD
ALA125825250MG ALA125825-25G ALA125825-50MG ALA125825-5G ALA125825-10MG ALA125825-1G
Atazanavir ≥98%, Moligand™
Atazanavir

Atazanavir(BMS 232632) is an highly potent HIV 1 protease inhibitor.A highly potent HIV-1 protease inhibitor

Formula: C₃₈H₅₂N₆O₇
MW: 704.87 g/mol
Melting Pt: 198.0 to 204.0 °C
Storage Temperature: Freezer
MDL Number: MFCD08435966
CAS Number: 198904-31-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44191038f8b8f55a293e3d0710ff3b949b5272c6&vid=44191038f8b8f55a293e3d0710ff3b949b5272c6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR