Tavaborole ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
AN-2690, 5-Fluorobenzo[c][1,2]oxaborol-1(3H)-ol
A125782-1G
A125782-250MG
ALA125782-1GEA
607.91
USD
ALA125782-1G
ALA125782250MG
Tavaborole ≥98%
Tavaborole
AN-2690(Tavaborole), an antifungal agent with activity against Trichophyton species, in a topical solution formulation, for the potential treatment of onychomycosis.
Formula:
C₇H₆BFO₂ MW: 151.93 g/mol Melting Pt: 132 °C Storage Temperature: Freezer |
MDL Number:
MFCD10699483 CAS Number: 174671-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...