Aliskiren ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (2S,4S,5S,7S)-5-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-2-isopropyl-7-(4-methoxy-3-(3-methoxypropoxy)benzyl)-8-methylnonanamide

A125767-10MG A125767-250MG A125767-50MG
ALA125767-10MGEA 123.41 USD
ALA125767-10MG ALA125767250MG ALA125767-50MG
Aliskiren ≥98%, Moligand™
Aliskiren

Aliskiren(CGP 60536) is a direct renin inhibitor with IC50 of 1.5 nM.

Formula: C₃₀H₅₃N₃O₆
MW: 551.75 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09839018
CAS Number: 173334-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def4e664438edc487b65e45847b8d9d13585e12f&vid=def4e664438edc487b65e45847b8d9d13585e12f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR