Piperidin-1-yl(quinoxalin-6-yl)methanone ≥95%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-(Piperidin-1-ylcarbonyl)quinoxaline, BDP 12, CX516, Ampalex, Ampakine CX516, 1-Piperidinyl-6-quinoxalinyl-methanone

A125747-100MG A125747-1G A125747-500MG
ALA125747100MGEA 148.11 USD
ALA125747100MG ALA125747-1G ALA125747500MG
Piperidin-1-yl(quinoxalin-6-yl)methanone ≥95%, Moligand™
Piperidin-1-yl(quinoxalin-6-yl)methanone

CX516 is a positive allosteric modulator at AMPA receptor that inhibits the deactivation of AMPA receptors. CX-516 is a nootropic and ampakine agentA series of AMPA modulators for the potential treatment of cognitive diseases

Formula: C₁₄H₁₅N₃O
Storage Temperature: Freezer
MDL Number: MFCD00943201
CAS Number: 154235-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35884c881005491bf0c57043117143a708e1cb97&vid=35884c881005491bf0c57043117143a708e1cb97&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR