AZD-3463 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A125664-100MG A125664-10MG A125664-25MG A125664-50MG A125664-5MG
ALA125664100MGEA 465.41 USD
ALA125664100MG ALA125664-10MG ALA125664-25MG ALA125664-50MG ALA125664-5MG
AZD-3463 ≥98%, Moligand™
AZD-3463

AZD3463 is a novel orally bioavailable ALK inhibitor with Ki of 0.75 nM, which also inhibits IGF1R with equivalent potency.

Formula: C₂₄H₂₅ClN₆O
Storage Temperature: Freezer
MDL Number: MFCD25976759
CAS Number: 1356962-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d442b3042c804f04e07b5b68345443b8db598200&vid=d442b3042c804f04e07b5b68345443b8db598200&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR