AZ20 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (3R)-4-[2-(3H-indol-4-yl)-6-(1-methylsulphonylcyclopropyl)pyrimidin-4-yl]-3-methylmorpholine, AZ20, AZ 20, (R)-4-(2-(1H-Indol-4-yl)-6-(1-(methylsulphonyl)cyclopropyl)pyrimidin-4-yl)-3-methylmorpholine

A125603-10MG A125603-50MG A125603-5MG
ALA125603-10MGEA 93.01 USD
ALA125603-10MG ALA125603-50MG ALA125603-5MG
AZ20 ≥98%, Moligand™
AZ20
Formula: C₂₁H₂₄N₄O₃S
MW: 412.51 g/mol
Storage Temperature: Freezer
MDL Number: MFCD25976730
CAS Number: 1233339-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc29c4704f36e27734dc6ec16eb0b1c1883456d7&vid=bc29c4704f36e27734dc6ec16eb0b1c1883456d7&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR