AS-703026 ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
(S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamide, MSC1936369B, Pimasertib
A125227-50MG
A125227-5MG
ALA125227-50MGEA
186.11
USD
ALA125227-50MG
ALA125227-5MG
AS-703026 ≥98%, Moligand™
AS-703026
Pimasertib (AS-703026) is a highly selective, potent, ATP non-competitive allosteric inhibitor of MEK1/2 with IC50 of 5 nM-2 μM in MM cell lines.A potent inhibitor of MEK1 and MEK2.
Formula:
C₁₅H₁₅FIN₃O₃ MW: 431.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16660676 CAS Number: 1236699-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...