AR-C155858 ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: (S)-6-((3,5-Dimethyl-1H-pyrazol-4-yl)methyl)-5-(4-hydroxyisoxazolidine-2-carbonyl)-1-isobutyl-3-methylthieno[2,3-d]pyrimidine-2,4(1H,3H)-dione, 6-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]-5-[[(4S)-4-hydroxy-2-isoxazolidinyl]carbonyl]-3-methyl-1-(2-methylpropyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione

A125208-10MG A125208-1MG A125208-25MG A125208-50MG A125208-5MG
ALA125208-10MGEA 586.06 USD
ALA125208-10MG ALA125208-1MG ALA125208-25MG ALA125208-50MG ALA125208-5MG
AR-C155858 ≥95%
AR-C155858
Formula: C₂₁H₂₇N₅O₅S
MW: 461.53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22124476
CAS Number: 496791-37-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6da0f69a6298dc28957f8446f50506dda1179cde&vid=6da0f69a6298dc28957f8446f50506dda1179cde&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR