AGI-6780 ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide

A124933-50MG A124933-5MG
ALA124933-50MGEA 470.16 USD
ALA124933-50MG ALA124933-5MG
AGI-6780 ≥98%
AGI-6780

AGI-6780 is a potent and selective inhibitor of IDH2 R140Q mutant with IC50 of 23 nM.

Formula: C₂₁H₁₈F₃N₃O₃S₂
MW: 481.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26097285
CAS Number: 1432660-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70831963be2ad5f9f14dcfacca41ea8cac9c454e&vid=70831963be2ad5f9f14dcfacca41ea8cac9c454e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR