ACY-1215 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Rocilinostat, Ricolinostat, 2-(diphenylamino)-N-(7-(hydroxyamino)-7-oxoheptyl)pyrimidine-5-carboxamide, ACT-1215

A124886-100MG A124886-10MG A124886-50MG A124886-5MG
ALA124886100MGEA 133.86 USD
ALA124886100MG ALA124886-10MG ALA124886-50MG ALA124886-5MG
ACY-1215 ≥98%, Moligand™
ACY-1215

Rocilinostat (ACY-1215) is a selective HDAC6 inhibitor with IC50 of 5 nM. It is >10-fold more selective for HDAC6 than HDAC1/2/3 (class I HDACs) with slight activity against HDAC8, minimal activity against HDAC4/5/7/9/11, Sirtuin1, and Sirtuin2.

Formula: C₂₄H₂₇N₅O₃
MW: 433.50 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22666356
CAS Number: 1316214-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=465e14a2dc94390e71b331bb2865cbb9b6f02260&vid=465e14a2dc94390e71b331bb2865cbb9b6f02260&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR