Aripiprazole ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 7-[4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy]-3,4-dihydro-2(1H)-quinolinone

A124880-1G A124880-25G A124880-5G
ALA124880-1GEA 167.11 USD
ALA124880-1G ALA124880-25G ALA124880-5G
Aripiprazole ≥99%, Moligand™
Aripiprazole

Aripiprazole is a human 5-HT1A receptor partial agonist with a Ki of 4.2 nMA selective dopamine D2-receptor antagonist showing autoreceptor agonist activity.Aripiprazole has been used to study behavioral and neurochemical effects of aripiprazole treatment in juvenile spontaneously hypertensive rats (SHR), a model for attention deficit hyperactivity disorder (ADHD).Aripiprazole has been used to study its effect on mammary tumorigenesis, mediated by retrovirus delivery of receptor tyrosine-protein kinase 2 or HRas proto-oncogene, GTPase or by transgenic expression of Wnt-1 (proto-oncoprotein). It has also been used to study its behavioral and neurochemical effects in juvenile spontaneously hypertensive rats.

Formula: C₂₃H₂₇Cl₂N₃O₂
MW: 448.39 g/mol
Melting Pt: 149 °C
Storage Temperature: Freezer
MDL Number: MFCD00892072
CAS Number: 129722-12-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57ba2536980a254d0731ff6d45abbe65d23b2312&vid=57ba2536980a254d0731ff6d45abbe65d23b2312&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR