7-Azaindole ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

A124841-100G A124841-25G A124841-5G
ALA124841-100GEA 325.76 USD
ALA124841-100G ALA124841-25G ALA124841-5G
7-Azaindole ≥98%
7-Azaindole

A heterocyclic molecule that can be utilized as a pharmaceutical building block, Starting material in a recent synthesis of azaserotonin.

Formula: C₇H₆N₂
Boiling Pt: 270°C
Melting Pt: 105-107°C
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f60c8bb95a1eafb746e24419dbfc831b370b1b5&vid=5f60c8bb95a1eafb746e24419dbfc831b370b1b5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR