Atenolol ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide

A124776-100G A124776-1G A124776-25G A124776-5G
ALA124776-100GEA 2447.11 USD
ALA124776-100G ALA124776-1G ALA124776-25G ALA124776-5G
Atenolol ≥98%, Moligand™
Atenolol

(RS)-Atenolol is a cardioselective β-adrenergic blocker. It is considered to be antihypertensive, antianginal, and antiarrhythmic. (RS)-Atenolol is an inhibitor of β1-AR and β2-AR.An inhibitor of β1-AR and β2-AR.Atenolol is a β-adrenergic blocker and used in the management of hypertension. Atenolol has antianginal and antiarrhythmic properties.

Formula: C₁₄H₂₂N₂O₃
MW: 266.34 g/mol
Melting Pt: 151-155°C
Storage Temperature: Freezer
MDL Number: MFCD00057645
CAS Number: 29122-68-7
EINECS: 249-451-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8324e47a3ad8bf5483d18b1304d9ee33efa14482&vid=8324e47a3ad8bf5483d18b1304d9ee33efa14482&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR