Allyltriphenylphosphonium bromide ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A124746-25G A124746-5G
ALA124746-25GEA 71.16 USD
ALA124746-25G ALA124746-5G
Allyltriphenylphosphonium bromide ≥98%
Allyltriphenylphosphonium bromide

Reactant for: Preparation of a diol and carbamate chemistry library for common functional groups Regioselective synthesis of alkenes via semihydrogenation and semihydrogenation-oxidation of dienes Alkene addition of frustrated Lewis pairs Olefination of N-sulfonyl imines for stereoselective synthesis of vinyl arenes Wittig olefination of aldehydes for preparation of conjugated dienes Tandem Michael addition / ylide olefination reactions for the synthesis of highly functionalized cyclohexadienes

Formula: C₂₁H₂₀BrP
MW: 383.261 g/mol
Melting Pt: 222-225°C
Storage Temperature: Ambient
MDL Number: MFCD00011808
CAS Number: 1560-54-9
EINECS: 216-332-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e4f2a5cbe08483d05a356c5ba9a5eec1243fd6c&vid=9e4f2a5cbe08483d05a356c5ba9a5eec1243fd6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR