3-Amino-5-tert-butylisoxazole ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
A124704-25G
A124704-5G
ALA124704-25GEA
173.76
USD
ALA124704-25G
ALA124704-5G
3-Amino-5-tert-butylisoxazole ≥97%
3-Amino-5-tert-butylisoxazole
Amination of aryl bromides catalyzed by Pd(dba)3 and Xantphos.
Formula:
C₇H₁₂N₂O MW: 140.19 g/mol Melting Pt: 110-113°C |
MDL Number:
MFCD00055620 CAS Number: 55809-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...