N-(2-Aminoethyl)glycine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A124646-1G A124646-250MG A124646-50MG
ALA124646-1GEA 444.51 USD
ALA124646-1G ALA124646250MG ALA124646-50MG
N-(2-Aminoethyl)glycine ≥97%
N-(2-Aminoethyl)glycine
Formula: C₄H₁₀N₂O₂
Melting Pt: 143°C
MDL Number: MFCD00144824
CAS Number: 24123-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ded2f11af167443192e9386f268f5cbe61debd75&vid=ded2f11af167443192e9386f268f5cbe61debd75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR