1-Boc-(2-aminomethyl)piperidine ≥95%

Supplier: Aladdin Scientific
Warning

A124599-1G A124599-250MG A124599-25G A124599-5G
ALA124599-1GEA 74.96 USD
ALA124599-1G ALA124599250MG ALA124599-25G ALA124599-5G
1-Boc-(2-aminomethyl)piperidine ≥95%
1-Boc-(2-aminomethyl)piperidine

Reactant for synthesis of: TRPV1 antagonists CHK1 inhibitors p38a MAP kinase inhibitors Carbamoylphosphonates Macrolactams via c

Formula: C₁₁H₂₂N₂O₂
Density: 1.0124 g/cm³
Flash Pt: >110°C
Storage Temperature: Refrigerator
MDL Number: MFCD01317791
CAS Number: 370069-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdd395426abd6593a065859558019d8cef8b450b&vid=cdd395426abd6593a065859558019d8cef8b450b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR