3-Aminoisoxazole ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A124591-25ML A124591-5ML
ALA124591-25MLEA 508.16 USD
ALA124591-25ML ALA124591-5ML
3-Aminoisoxazole ≥95%
3-Aminoisoxazole

It is a 3-substituted isoxazole derivative. It is a structural isomer of 5-aminoisoxazole.It may be used in the following studies: . As a reagent in the synthesis of N-(4-(N-isoxazol-3-ylsulfamoyl)phenyl)acetamide. . As a starting material in the synthesis of N-(isoxazol-3-yl)-N’-(carbomethoxy)thiourea. . As a starting material in the synthesis of (Z)-2-(5-amino-1,2,4-thiadiazol-3-yl)-2-methoxy-iminoacetic acid, a side-chain of the fourth generation of cephem antibiotics.

Formula: C₃H₄N₂O
Boiling Pt: 226-228°C
Density: 1.138 g/cm³
Flash Pt: 113 °C
MDL Number: MFCD00038814
CAS Number: 1750-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0728c60445128b258ca3758334acc9b9b548cbe9&vid=0728c60445128b258ca3758334acc9b9b548cbe9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR