1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline, 4-(4-(4-Methoxyphenyl)piperazin-1-yl)aniline

A124582-1G A124582-25G
ALA124582-1GEA 9.41 USD
ALA124582-1G ALA124582-25G
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96%
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Formula: C₁₇H₂₁N₃O
Melting Pt: 186°C
MDL Number: MFCD02071496
CAS Number: 74852-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f66cc7a221416af79ed0057f7c76e61aa109094&vid=4f66cc7a221416af79ed0057f7c76e61aa109094&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR